Assay Details
Target Gene Details
Entrez Gene ID: | 84674 |
Gene Name: | caspase recruitment domain family member 6 |
Gene Aliases: |
CINCIN1 |
Location: |
Chr.5:40841190-40855354 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARD6 | NM_032587.3 | 3 | 1586 | NP_115976.2 |
| XM_017009989.1 | 2 | 1139 | XP_016865478.1 | |
| XM_017009990.1 | 4 | 1820 | XP_016865479.1 | |
| AB209844.1 | 3 | 1406 | BAD93081.1 | |
| AF356193.1 | 3 | 1710 | AAK32718.1 | |
| AK304349.1 | ||||
| AY196783.1 | AAP40637.1 | |||
| BC093825.1 | 3 | 1533 | AAH93825.1 |
More Information
Set Membership: |
|

Genomic Map