Assay Details
Target Gene Details
Entrez Gene ID: | 9026 |
Gene Name: | huntingtin interacting protein 1 related |
Gene Aliases: |
HIP12, HIP3, ILWEQ |
Location: |
Chr.12:122834498-122862961 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIP1R | NM_001303097.1 | NP_001290026.1 | ||
| NM_001303099.1 | NP_001290028.1 | |||
| NM_003959.2 | NP_003950.1 | |||
| XM_011538963.1 | XP_011537265.1 | |||
| AB013384.1 | BAA33713.1 | |||
| AB014555.1 | ||||
| AK090537.1 | ||||
| AK298387.1 | ||||
| BC067085.1 | AAH67085.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560490 | Chr.12:122841827 - 122862424 on Build GRCh38 | Loss |
|
| nsv952498 | Chr.12:122839654 - 122868353 on Build GRCh38 | Deletion |
|
| nsv517200 | Chr.12:122827504 - 123169951 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map