Assay Details
Target Gene Details
Entrez Gene ID: | 260293 |
Gene Name: | cytochrome P450 family 4 subfamily X member 1 |
Gene Aliases: |
CYPIVX1 |
Location: |
Chr.1:46961364-47055432 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CYP4X1 | NM_001320289.1 | 2 | 454 | NP_001307218.1 |
| NM_001320290.1 | 2 | 191 | NP_001307219.1 | |
| NM_178033.1 | 2 | 488 | NP_828847.1 | |
| XM_017000973.1 | 2 | 519 | XP_016856462.1 | |
| AK091806.1 | 2 | 192 | BAC03751.1 | |
| AK098065.1 | 2 | 481 | BAC05226.1 | |
| AK131355.1 | 2 | 454 | BAD18508.1 | |
| AM040940.1 | 2 | 325 | CAJ13826.1 | |
| AY358537.1 | 2 | 471 | AAQ88901.1 | |
| BC028102.1 | 2 | 488 | AAH28102.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546167 | Chr.1:47021913 - 47058592 on Build GRCh38 | Gain |
|
| esv2763300 | Chr.1:46892860 - 47163102 on Build GRCh38 | Gain |
|
| esv3585949 | Chr.1:47009618 - 47041416 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map