Assay Details
Target Gene Details
Entrez Gene ID: | 4155 |
Gene Name: | myelin basic protein |
Gene Aliases: |
- |
Location: |
Chr.18:76978833-77133708 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MBP | NM_001025081.1 | 7 | 825 | NP_001020252.1 |
| NM_001025090.1 | 6 | 747 | NP_001020261.1 | |
| NM_001025092.1 | 5 | 714 | NP_001020263.1 | |
| NM_001025101.1 | 9 | 1319 | NP_001020272.1 | |
| NM_002385.2 | 6 | 792 | NP_002376.1 | |
| AK094611.1 | 1 | 911 | ||
| AK095121.1 | 1 | 1701 | ||
| AK098402.1 | 6 | 703 | ||
| AK122594.1 | 2 | 1270 | ||
| AK123433.1 | 2 | 3367 | BAC85616.1 | |
| AK124830.1 | 2 | 2044 | BAC85961.1 | |
| AK126858.1 | 1 | 3861 | ||
| AK128770.1 | 6 | 706 | ||
| AK128788.1 | 6 | 703 | ||
| AK289893.1 | 6 | 747 | ||
| AK293922.1 | 5 | 742 | ||
| BC008749.2 | 6 | 685 | ||
| BC080654.1 | 5 | 659 | AAH80654.1 | |
| BG766942.1 | 6 | 746 | ||
| CB156561.1 | ||||
| CR627018.1 | 4 | 8477 | CAH10359.1 | |
| M13577.1 | 6 | 692 | AAA59562.1 | |
| M30047.1 | 5 | 655 | AAA59559.1 | |
| M30515.1 | 8 | 741 | AAA59564.1 | |
| M30516.1 | 6 | 711 | AAA59563.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2717388 | Chr.18:76288671 - 78160297 on Build GRCh38 | Deletion |
|
| esv3643197 | Chr.18:76911767 - 77002929 on Build GRCh38 | Loss |
|
| nsv1063632 | Chr.18:76317786 - 77170410 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map