Assay Details
Target Gene Details
Entrez Gene ID: | 388697 |
Gene Name: | hornerin |
Gene Aliases: |
FLG3, S100A16, S100a18 |
Location: |
Chr.1:152212076-152224196 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HRNR | NM_001009931.2 | 3 | 8762 | NP_001009931.1 |
AB104446.1 | 1 | 1612 | BAC57496.1 | |
AY396741.1 | 3 | 8762 | AAR91619.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1014095 | Chr.1:152132010 - 152482115 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv8391 | Chr.1:152212852 - 152221607 on Build GRCh38 | Gain+Loss |
![]() |
nsv514913 | Chr.1:152212884 - 152214676 on Build GRCh38 | Gain |
![]() |
nsv1007097 | Chr.1:151860320 - 152255501 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv57 | Chr.1:152210699 - 152214675 on Build GRCh38 | Insertion |
![]() |
nsv509468 | Chr.1:152187656 - 152299397 on Build GRCh38 | Insertion |
![]() |
nsv2877 | Chr.1:152191047 - 152233114 on Build GRCh38 | Insertion |
![]() |
nsv471347 | Chr.1:152212082 - 152224193 on Build GRCh38 | Gain |
![]() |
nsv547853 | Chr.1:152036195 - 152264284 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
