Assay Details
Target Gene Details
Entrez Gene ID: | 1296 |
Gene Name: | collagen type VIII alpha 2 |
Gene Aliases: |
FECD, FECD1, PPCD, PPCD2 |
Location: |
Chr.1:36095236-36126207 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COL8A2 | NM_001294347.1 | 4 | 1973 | NP_001281276.1 |
| NM_005202.3 | 4 | 2118 | NP_005193.1 | |
| XM_005270477.3 | 4 | 2818 | XP_005270534.1 | |
| AK074129.1 | 2 | 2036 | ||
| BC096295.1 | 2 | 691 | AAH96295.1 | |
| BC096296.3 | 3 | 1208 | AAH96296.1 | |
| BC096297.1 | 2 | 1891 | AAH96297.1 | |
| BC099645.1 | 2 | 1891 | AAH99645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834691 | Chr.1:36059137 - 36238540 on Build GRCh38 | Loss |
|
| nsv545992 | Chr.1:36091266 - 36161590 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map