Assay Details
Target Gene Details
Entrez Gene ID: | 222698 |
Gene Name: | NFKB activating protein like |
Gene Aliases: |
C6orf194, bA424I5.1 |
Location: |
Chr.6:28259297-28260958 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NKAPL | NM_001007531.2 | 1 | 42 | NP_001007532.1 |
| BC038240.1 | AAH38240.1 | |||
| BC101681.1 | AAI01682.1 | |||
| BC101685.1 | AAI01686.1 | |||
| DB471905.1 | 1 | 42 |
Target Gene Details
Entrez Gene ID: | 387032 |
Gene Name: | zinc finger with KRAB and SCAN domains 4 |
Gene Aliases: |
P1P373C6, ZNF307, ZNF427, ZSCAN36 |
Location: |
Chr.6:28241697-28259312 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZKSCAN4 | XM_005249100.4 | XP_005249157.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv950146 | Chr.6:28259123 - 28267322 on Build GRCh38 | Duplication |
|
| nsv830620 | Chr.6:28206219 - 28380256 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map