Assay Details
Target Gene Details
Entrez Gene ID: | 64081 |
Gene Name: | phenazine biosynthesis like protein domain containing |
Gene Aliases: |
HEL-S-306, MAWBP, MAWDBP |
Location: |
Chr.10:68282660-68332958 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PBLD | NM_022129.3 | 10 | 1983 | NP_071412.2 |
| XM_005270028.3 | 10 | 2017 | XP_005270085.1 | |
| XM_011540060.2 | 10 | 1992 | XP_011538362.1 | |
| XM_017016513.1 | 10 | 1951 | XP_016872002.1 | |
| AB049758.1 | 8 | 1845 | BAB16606.1 | |
| AK027673.1 | 10 | 1950 | BAB55285.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3623705 | Chr.10:68099008 - 68297411 on Build GRCh38 | Gain |
|
| esv3623707 | Chr.10:68214112 - 68304911 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map