Assay Details
Target Gene Details
Entrez Gene ID: | 5463 |
Gene Name: | POU class 6 homeobox 1 |
Gene Aliases: |
BRN5, MPOU, TCFB1 |
Location: |
Chr.12:51186936-51218134 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU6F1 | NR_026893.1 | |||
| XM_017019507.1 | 5 | 1686 | XP_016874996.1 | |
| XM_017019508.1 | 6 | 1597 | XP_016874997.1 | |
| XM_017019509.1 | 4 | 606 | XP_016874998.1 | |
| XM_017019510.1 | 5 | 1035 | XP_016874999.1 | |
| XM_017019511.1 | 5 | 1204 | XP_016875000.1 | |
| XM_017019512.1 | 5 | 560 | XP_016875001.1 | |
| XM_017019513.1 | 5 | 1686 | XP_016875002.1 | |
| XM_017019514.1 | 5 | 1686 | XP_016875003.1 | |
| XM_017019515.1 | 5 | 1032 | XP_016875004.1 | |
| XM_017019516.1 | 5 | 1686 | XP_016875005.1 | |
| XM_017019517.1 | 5 | 1686 | XP_016875006.1 | |
| XM_017019518.1 | 5 | 1686 | XP_016875007.1 | |
| XM_017019519.1 | 5 | 1686 | XP_016875008.1 | |
| XM_017019520.1 | XP_016875009.1 | |||
| XM_017019521.1 | XP_016875010.1 | |||
| XM_017019522.1 | XP_016875011.1 | |||
| XM_017019523.1 | XP_016875012.1 | |||
| XM_017019524.1 | XP_016875013.1 | |||
| XM_017019525.1 | XP_016875014.1 | |||
| XM_017019526.1 | XP_016875015.1 | |||
| XM_017019527.1 | XP_016875016.1 | |||
| XM_017019528.1 | XP_016875017.1 | |||
| XM_017019529.1 | XP_016875018.1 | |||
| XM_017019530.1 | XP_016875019.1 | |||
| AK095191.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34039 | Chr.12:50932919 - 51278127 on Build GRCh38 | Loss |
|
| nsv558876 | Chr.12:51197584 - 51218946 on Build GRCh38 | Gain |
|
More Information
Additional Information:
Set Membership: |
|

Genomic Map