Assay Details
Target Gene Details
Entrez Gene ID: | 22861 |
Gene Name: | NLR family pyrin domain containing 1 |
Gene Aliases: |
CARD7, CIDED, CLR17.1, DEFCAP, DEFCAP-L/S, NAC, NALP1, PP1044, SLEV1, VAMAS1 |
Location: |
Chr.17:5501399-5584512 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP1 | NM_001033053.2 | NP_001028225.1 | ||
| AK026398.1 | BAB15470.1 | |||
| BP391071.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1956 | Chr.17:5470460 - 5509582 on Build GRCh38 | Deletion |
|
| dgv884e212 | Chr.17:5496610 - 5516416 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map