Assay Details
Target Gene Details
Entrez Gene ID: | 11065 |
Gene Name: | ubiquitin conjugating enzyme E2 C |
Gene Aliases: |
UBCH10, dJ447F3.2 |
Location: |
Chr.20:45812576-45816957 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2C | NM_001281741.1 | NP_001268670.1 | ||
| NM_001281742.1 | NP_001268671.1 | |||
| NM_007019.3 | NP_008950.1 | |||
| NM_181799.2 | NP_861515.1 | |||
| NM_181800.2 | NP_861516.1 | |||
| NR_104036.1 | ||||
| NR_104037.1 | ||||
| AK311949.1 | ||||
| BC007656.2 | AAH07656.1 | |||
| BC016292.1 | AAH16292.1 | |||
| BC050736.1 | AAH50736.1 | |||
| BM556795.1 | ||||
| BM747513.1 | ||||
| BT007300.1 | AAP35964.1 | |||
| BT020165.1 | AAV38967.1 | |||
| BT020166.1 | AAV38968.1 | |||
| BU169315.1 | ||||
| BU171488.1 | ||||
| BU844974.1 | ||||
| CB143441.1 | ||||
| CK904708.1 | ||||
| CN354813.1 | ||||
| CN354815.1 | ||||
| CR456988.1 | CAG33269.1 | |||
| DB062573.1 | ||||
| U73379.1 | AAB53362.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833990 | Chr.20:45771510 - 45946844 on Build GRCh38 | Loss |
|
| nsv586132 | Chr.20:45785138 - 45842587 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map