Assay Details
Target Gene Details
Entrez Gene ID: | 83876 |
Gene Name: | maestro |
Gene Aliases: |
B29, C18orf3 |
Location: |
Chr.18:50795120-50825413 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRO | NM_001127174.1 | NP_001120646.1 | ||
| NM_001127175.1 | 1 | 103 | NP_001120647.1 | |
| NM_001127176.1 | 1 | 103 | NP_001120648.1 | |
| NM_031939.3 | 1 | 103 | NP_114145.2 | |
| XM_005258365.3 | XP_005258422.1 | |||
| XM_011526226.2 | XP_011524528.1 | |||
| XM_017026036.1 | XP_016881525.1 | |||
| XM_017026037.1 | 1 | 151 | XP_016881526.1 | |
| AB042647.1 | BAB40981.1 | |||
| AK054702.1 | BAB70797.1 | |||
| AK295666.1 | 1 | 104 | ||
| AL705398.1 | ||||
| BC029860.1 | AAH29860.1 | |||
| DC333169.1 | 1 | 104 | ||
| DC403805.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527714 | Chr.18:50818407 - 50888556 on Build GRCh38 | Gain |
|
| nsv9630 | Chr.18:50532107 - 50922466 on Build GRCh38 | Loss |
|
| nsv576892 | Chr.18:50746815 - 50899947 on Build GRCh38 | Gain |
|

Genomic Map