Assay Details
Target Gene Details
Entrez Gene ID: | 50717 |
Gene Name: | DDB1 and CUL4 associated factor 8 |
Gene Aliases: |
GAN2, H326, WDR42A |
Location: |
Chr.1:160215715-160262560 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF8 | NM_015726.3 | 14 | 2083 | NP_056541.2 |
| NR_028103.1 | 14 | 2383 | ||
| NR_028104.1 | 13 | 2309 | ||
| AK026481.1 | ||||
| AK223475.1 | 13 | 1938 | BAD97195.1 | |
| AK303188.1 | ||||
| BC098271.1 | 14 | 1989 | AAH98271.1 | |
| BC099709.1 | 14 | 1989 | AAH99709.1 | |
| BC099846.1 | 14 | 1989 | AAH99846.1 | |
| CR749801.1 | 14 | 2306 | CAH18661.1 | |
| U06631.1 | 14 | 2047 | AAA16607.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76573195] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map