Assay Details
Target Gene Details
Entrez Gene ID: | 3671 |
Gene Name: | immunoglobulin superfamily containing leucine rich repeat |
Gene Aliases: |
HsT17563, Meflin |
Location: |
Chr.15:74173746-74176871 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ISLR | NM_005545.3 | 1 | 210 | NP_005536.1 |
| AB024536.1 | 1 | 210 | BAA85970.1 | |
| AK000817.1 | 1 | 210 | ||
| AK074668.1 | 1 | 618 | ||
| AK297730.1 | ||||
| AY358871.1 | 1 | 286 | AAQ89230.1 | |
| BC111013.1 | 1 | 120 | AAI11014.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833053 | Chr.15:74067815 - 74262799 on Build GRCh38 | Loss |
|
| nsv1050884 | Chr.15:74102642 - 74197072 on Build GRCh38 | Gain |
|
| nsv469619 | Chr.15:73999518 - 74173980 on Build GRCh38 | Loss |
|
| nsv833054 | Chr.15:74165302 - 74332942 on Build GRCh38 | Loss |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map