Assay Details
Target Gene Details
Entrez Gene ID: | 25764 |
Gene Name: | huntingtin interacting protein K |
Gene Aliases: |
C15orf63, HSPC136 |
Location: |
Chr.15:43800421-43802589 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HYPK | NM_001199885.1 | 3 | 822 | NP_001186814.1 |
| NM_016400.3 | 4 | 878 | NP_057484.3 | |
| AK022435.1 | 2 | 785 | ||
| BC019262.1 | 4 | 876 | AAH19262.2 |
Target Gene Details
Entrez Gene ID: | 100529067 |
Gene Name: | SERF2-C15orf63 readthrough |
Gene Aliases: |
- |
Location: |
Chr.15:43791976-43802589 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERF2-C15ORF63 | NR_037673.1 | 6 | 1321 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1036318 | Chr.15:43761419 - 43868575 on Build GRCh38 | Gain |
|
| esv2760028 | Chr.15:43530960 - 43856443 on Build GRCh38 | Gain+Loss |
|
| nsv1050263 | Chr.15:43801729 - 43911849 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map