Assay Details
Target Gene Details
Entrez Gene ID: | 57333 |
Gene Name: | reticulocalbin 3 |
Gene Aliases: |
RLP49 |
Location: |
Chr.19:49527579-49543638 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RCN3 | NM_020650.2 | NP_065701.2 | ||
| XM_005259089.3 | XP_005259146.1 | |||
| XM_011527143.1 | XP_011525445.1 | |||
| XM_017027023.1 | XP_016882512.1 | |||
| AF183423.1 | AAG09692.1 | |||
| AY195859.1 | AAO43054.1 | |||
| AY358423.1 | AAQ88789.1 | |||
| BC013436.1 | AAH13436.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9739 | Chr.19:47901840 - 50143063 on Build GRCh38 | Gain+Loss |
|
| nsv833863 | Chr.19:49479177 - 49629716 on Build GRCh38 | Loss |
|
| nsv833862 | Chr.19:49448789 - 49570676 on Build GRCh38 | Loss |
|
| nsv1160655 | Chr.19:49489110 - 49841830 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77227069] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map