Assay Details
Target Gene Details
Entrez Gene ID: | 5929 |
Gene Name: | RB binding protein 5, histone lysine methyltransferase complex subunit |
Gene Aliases: |
RBQ3, SWD1 |
Location: |
Chr.1:205086142-205122022 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP5 | NM_001193272.1 | NP_001180201.1 | ||
| NM_001193273.1 | NP_001180202.1 | |||
| NM_005057.3 | NP_005048.2 | |||
| XM_011509833.2 | XP_011508135.1 | |||
| AK290137.1 | ||||
| AK296978.1 | ||||
| AK297540.1 | ||||
| BC037284.2 | AAH37284.1 | |||
| BC053856.1 | AAH53856.1 | |||
| BC075059.2 | AAH75059.1 | |||
| BC075060.2 | AAH75060.1 | |||
| X85134.1 | CAA59446.1 |
More Information
Set Membership: |
|

Genomic Map