Assay Details
Target Gene Details
Entrez Gene ID: | 7566 |
Gene Name: | zinc finger protein 18 |
Gene Aliases: |
HDSG1, KOX11, ZKSCAN6, ZNF535, ZSCAN38, Zfp535 |
Location: |
Chr.17:11957433-11997510 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF18 | NM_001303281.1 | 7 | 1452 | NP_001290210.1 |
| NM_001303282.1 | 7 | 1449 | NP_001290211.1 | |
| NM_144680.3 | 9 | 1895 | NP_653281.2 | |
| XM_011524002.2 | 7 | 1525 | XP_011522304.1 | |
| XM_017025006.1 | 7 | 1452 | XP_016880495.1 | |
| XM_017025007.1 | 8 | 1795 | XP_016880496.1 | |
| XM_017025008.1 | 7 | 1449 | XP_016880497.1 | |
| XM_017025009.1 | 6 | 1777 | XP_016880498.1 | |
| XM_017025010.1 | 7 | 1522 | XP_016880499.1 | |
| XM_017025011.1 | 8 | 1792 | XP_016880500.1 | |
| AK057783.1 | BAB71570.1 | |||
| AK092656.1 | 9 | 1895 | BAC03939.1 | |
| AK127908.1 | 7 | 1410 | ||
| AY491053.1 | 7 | 1396 | AAS67595.1 | |
| BC036096.2 | 6 | 1373 | AAH36096.1 | |
| X52342.1 | CAA36568.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639962 | Chr.17:11973639 - 11978532 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map