Assay Details
Target Gene Details
Entrez Gene ID: | 29888 |
Gene Name: | striatin 4 |
Gene Aliases: |
PPP2R6C, ZIN, zinedin |
Location: |
Chr.19:46719509-46746475 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRN4 | NM_001039877.1 | 10 | 1434 | NP_001034966.1 |
| NM_013403.2 | 10 | 1413 | NP_037535.2 | |
| XM_006723171.2 | 11 | 1491 | XP_006723234.1 | |
| XM_006723172.2 | 11 | 1470 | XP_006723235.1 | |
| XM_006723174.2 | 10 | 1241 | XP_006723237.1 | |
| XM_011526878.1 | 11 | 1615 | XP_011525180.1 | |
| XM_011526879.1 | 4 | 578 | XP_011525181.1 | |
| XM_017026716.1 | 11 | 1220 | XP_016882205.1 | |
| XM_017026717.1 | 10 | 1196 | XP_016882206.1 | |
| XM_017026718.1 | 6 | 985 | XP_016882207.1 | |
| XM_017026719.1 | 10 | 1570 | XP_016882208.1 | |
| AB209003.1 | 10 | 1386 | BAD92240.1 | |
| AF212940.1 | 10 | 1380 | AAF29527.1 | |
| AK056294.1 | 4 | 435 | ||
| AK092913.1 | 1 | 749 | ||
| AK225493.1 | 4 | 409 | ||
| AK297637.1 | 11 | 1392 | ||
| AK298804.1 | 11 | 1460 | ||
| AK308813.1 | 5 | 648 | ||
| BC004910.2 | 2 | 168 | AAH04910.2 | |
| BC034604.1 | 10 | 1390 | AAH34604.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833850 | Chr.19:46703285 - 46867053 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map