Assay Details
Target Gene Details
Entrez Gene ID: | 56963 |
Gene Name: | repulsive guidance molecule family member a |
Gene Aliases: |
RGM |
Location: |
Chr.15:93043406-93089214 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RGMA | NM_001166283.1 | 3 | 822 | NP_001159755.1 |
| NM_001166286.1 | 2 | 682 | NP_001159758.1 | |
| NM_001166287.1 | 3 | 796 | NP_001159759.1 | |
| NM_001166288.1 | 3 | 680 | NP_001159760.1 | |
| NM_001166289.1 | 3 | 700 | NP_001159761.1 | |
| NM_020211.2 | 3 | 849 | NP_064596.2 | |
| AK074910.1 | 3 | 839 | BAC11285.1 | |
| AK074966.1 | 2 | 656 | BAC11321.1 | |
| AK074980.1 | 3 | 616 | BAC11330.1 | |
| AK125204.1 | 4 | 805 | ||
| AK291365.1 | 2 | 479 | ||
| AK299363.1 | 3 | 698 | ||
| AK309814.1 | 4 | 1062 | ||
| AL136826.1 | 3 | 663 | CAB66760.1 | |
| AL390083.1 | 2 | 605 | CAB98207.1 | |
| BC015886.1 | 2 | 673 | ||
| BC140838.1 | 3 | 627 | ||
| BC151132.1 | 3 | 627 | ||
| DA512691.1 | 3 | 680 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1052724 | Chr.15:93026471 - 93079437 on Build GRCh38 | Gain |
|
| nsv523617 | Chr.15:93027493 - 93066910 on Build GRCh38 | Loss |
|
| nsv1045427 | Chr.15:93033752 - 93055210 on Build GRCh38 | Gain |
|
| nsv833096 | Chr.15:93041512 - 93223028 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map