Assay Details
Target Gene Details
Entrez Gene ID: | 215 |
Gene Name: | ATP binding cassette subfamily D member 1 |
Gene Aliases: |
ABC42, ALD, ALDP, AMN |
Location: |
Chr.X:153724851-153744762 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCD1 | NM_000033.3 | 1 | 159 | NP_000024.2 |
| BC015541.1 | 1 | 140 | AAH15541.1 | |
| BC025358.1 | 1 | 159 | AAH25358.1 | |
| Z21876.1 | 1 | 146 | CAA79922.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
| esv21656 | Chr.X:153687433 - 153750282 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map