Assay Details
Target Gene Details
Entrez Gene ID: | 10241 |
Gene Name: | calcium binding and coiled-coil domain 2 |
Gene Aliases: |
NDP52 |
Location: |
Chr.17:48830988-48865245 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CALCOCO2 | NM_001261390.1 | 6 | 641 | NP_001248319.1 |
| NM_001261391.1 | 6 | 632 | NP_001248320.1 | |
| NM_001261393.1 | NP_001248322.1 | |||
| NM_001261395.1 | 4 | 379 | NP_001248324.1 | |
| NM_005831.4 | 5 | 569 | NP_005822.1 | |
| AK222666.1 | 5 | 519 | BAD96386.1 | |
| AK223227.1 | 5 | 512 | BAD96947.1 | |
| AK293137.1 | 4 | 332 | ||
| AK293329.1 | 6 | 594 | ||
| AK298177.1 | 6 | 584 | ||
| AK303313.1 | ||||
| AK304024.1 | 4 | 677 | ||
| AK314796.1 | 5 | 522 | ||
| AL042638.1 | 5 | 568 | ||
| BC004130.2 | 5 | 508 | AAH04130.1 | |
| BC015893.2 | 4 | 478 | AAH15893.1 | |
| BG773301.1 | 5 | 551 | ||
| CR456763.1 | 4 | 468 | CAG33044.1 | |
| U22897.1 | 5 | 522 | AAA75297.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482969 | Chr.17:48843507 - 48994527 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map