Assay Details
Target Gene Details
Entrez Gene ID: | 79935 |
Gene Name: | cyclin N-terminal domain containing 2 |
Gene Aliases: |
CCNP |
Location: |
Chr.19:40222208-40227111 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTD2 | NM_024877.3 | 5 | 1476 | NP_079153.2 |
| XM_006723395.3 | XP_006723458.2 | |||
| AI924992.1 | 1 | 343 | ||
| AK023327.1 | 4 | 1449 | BAB14529.1 | |
| BM709160.1 | ||||
| BU732215.1 | 1 | 353 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644345 | Chr.19:40203124 - 40288712 on Build GRCh38 | Gain |
|
| esv3644344 | Chr.19:40191059 - 40328585 on Build GRCh38 | Gain |
|
| esv2758761 | Chr.19:40106513 - 40333613 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map