Assay Details
Target Gene Details
Entrez Gene ID: | 782 |
Gene Name: | calcium voltage-gated channel auxiliary subunit beta 1 |
Gene Aliases: |
CAB1, CACNLB1, CCHLB1 |
Location: |
Chr.17:39173456-39197703 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CACNB1 | NM_000723.4 | NP_000714.3 | ||
| BC037311.1 | AAH37311.2 | |||
| M92303.1 | AAA35633.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833440 | Chr.17:39033205 - 39210437 on Build GRCh38 | Loss |
|
| esv3893012 | Chr.17:39172813 - 39177990 on Build GRCh38 | Loss |
|
| nsv833441 | Chr.17:39164554 - 39320543 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map