Assay Details
Target Gene Details
Entrez Gene ID: | 2200 |
Gene Name: | fibrillin 1 |
Gene Aliases: |
ACMICD, ECTOL1, FBN, GPHYSD2, MASS, MFLS, MFS1, OCTD, SGS, SSKS, WMS, WMS2 |
Location: |
Chr.15:48408306-48645788 on Build GRCh38 |
Assay Gene Location: | Within Exon 66 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FBN1 | NM_000138.4 | 66 | 8834 | NP_000129.3 |
| AB208840.1 | 30 | 3806 | BAD92077.1 | |
| AK309616.1 | 7 | 983 | ||
| BC146854.1 | 66 | 8700 | ||
| L13923.1 | 65 | 8572 | AAB02036.1 | |
| X63556.1 | 66 | 8834 | CAA45118.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047597 | Chr.15:48345836 - 48456074 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map