Assay Details
Target Gene Details
Entrez Gene ID: | 100507291 |
Gene Name: | uncharacterized LOC100507291 |
Gene Aliases: |
- |
Location: |
Chr.3:139389803-139583319 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507291 | NR_121609.1 | |||
| DA364121.1 |
Target Gene Details
Entrez Gene ID: | 5947 |
Gene Name: | retinol binding protein 1 |
Gene Aliases: |
CRABP-I, CRBP, CRBP1, CRBPI, RBPC |
Location: |
Chr.3:139517434-139539829 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBP1 | NM_002899.3 | NP_002890.2 | ||
| AK290315.1 | ||||
| BC121052.2 | ||||
| BU734286.1 | ||||
| CR541979.1 | CAG46776.1 | |||
| CR542005.1 | CAG46802.1 | |||
| M11433.1 | AAA60257.1 | |||
| M36809.1 | AAA35714.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001460 | Chr.3:139475791 - 139522774 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map