Assay Details
Target Gene Details
Entrez Gene ID: | 93380 |
Gene Name: | membrane magnesium transporter 1 |
Gene Aliases: |
EMC5, TMEM32 |
Location: |
Chr.X:135962072-135973975 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMGT1 | NM_173470.1 | 1 | 75 | NP_775741.1 |
| AK295829.1 | ||||
| AK312409.1 | 1 | 88 | ||
| AL157477.1 | 1 | 74 | ||
| BC033588.1 | 1 | 75 | AAH33588.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953696 | Chr.X:135973842 - 135974541 on Build GRCh38 | Deletion |
|
| esv2758895 | Chr.X:135468612 - 136072582 on Build GRCh38 | Gain+Loss |
|
| nsv1076276 | Chr.X:135973540 - 135976741 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map