Assay Details
Target Gene Details
Entrez Gene ID: | 134391 |
Gene Name: | G protein-coupled receptor 151 |
Gene Aliases: |
GALR4, GALRL, GPCR, PGR7 |
Location: |
Chr.5:146514854-146516113 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR151 | NM_194251.2 | 1 | 471 | NP_919227.2 |
| AY255557.1 | 1 | 84 | AAO85069.1 | |
| AY351676.1 | 1 | 471 | AAQ62567.1 |
More Information
Set Membership: |
|

Genomic Map