Assay Details
Target Gene Details
Entrez Gene ID: | 197257 |
Gene Name: | lactate dehydrogenase D |
Gene Aliases: |
DLD |
Location: |
Chr.16:75111860-75116772 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LDHD | NM_153486.3 | NP_705690.2 | ||
| NM_194436.2 | NP_919417.1 | |||
| AY092767.1 | AAM50322.1 | |||
| BC040279.1 | AAH40279.1 | |||
| BC047902.1 | AAH47902.1 | |||
| BM544922.1 |
More Information
Set Membership: |
|

Genomic Map