Assay Details
Target Gene Details
Entrez Gene ID: | 1911 |
Gene Name: | polyhomeotic homolog 1 |
Gene Aliases: |
EDR1, HPH1, MCPH11, RAE28 |
Location: |
Chr.12:8914664-8941467 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHC1 | XM_005253334.2 | XP_005253391.1 | ||
| XM_011520599.2 | XP_011518901.1 | |||
| XM_011520600.2 | XP_011518902.1 | |||
| XM_011520603.2 | XP_011518905.1 | |||
| XM_017018955.1 | XP_016874444.1 | |||
| XM_017018956.1 | XP_016874445.1 | |||
| XM_017018957.1 | XP_016874446.1 | |||
| XM_017018958.1 | XP_016874447.1 | |||
| AB209063.1 | BAD92300.1 | |||
| AK293893.1 | ||||
| BX640639.1 | CAE45787.1 | |||
| CN388285.1 | ||||
| U89277.1 | AAC51169.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051530 | Chr.12:8442286 - 9951690 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map