Assay Details
Target Gene Details
Entrez Gene ID: | 6277 |
Gene Name: | S100 calcium binding protein A6 |
Gene Aliases: |
2A9, 5B10, CABP, CACY, PRA |
Location: |
Chr.1:153534599-153536241 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A6 | NM_014624.3 | NP_055439.1 | ||
| XM_017002033.1 | XP_016857522.1 | |||
| AK312088.1 | ||||
| BC001431.2 | AAH01431.1 | |||
| BC009017.1 | AAH09017.1 | |||
| BF726732.1 | ||||
| BT006965.1 | AAP35611.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831603 | Chr.1:153433366 - 153603667 on Build GRCh38 | Gain |
|
| nsv1002705 | Chr.1:153335376 - 153553113 on Build GRCh38 | Gain |
|
| esv3587591 | Chr.1:153528630 - 153570193 on Build GRCh38 | Gain |
|
| esv3587584 | Chr.1:153466126 - 153598991 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map