Assay Details
Target Gene Details
Entrez Gene ID: | 10039 |
Gene Name: | poly(ADP-ribose) polymerase family member 3 |
Gene Aliases: |
ADPRT3, ADPRTL2, ADPRTL3, ARTD3, IRT1, PADPRT-3 |
Location: |
Chr.3:51942282-51948867 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP3 | NM_001003931.3 | NP_001003931.3 | ||
| NM_005485.5 | NP_005476.4 | |||
| XM_005264779.4 | XP_005264836.2 | |||
| XM_017005490.1 | XP_016860979.1 | |||
| AB208803.1 | BAD92040.1 | |||
| AF083068.1 | AAD29855.1 | |||
| AL050034.1 | CAB43246.1 | |||
| AY126341.1 | AAM95460.1 | |||
| BC014260.2 | AAH14260.1 | |||
| Y16836.1 | CAC79988.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1003453 | Chr.3:51928065 - 52065425 on Build GRCh38 | Gain |
|
| nsv508922 | Chr.3:51940932 - 52009520 on Build GRCh38 | Insertion |
|
| nsv954857 | Chr.3:51932485 - 52056584 on Build GRCh38 | Deletion |
|
| nsv520211 | Chr.3:51738331 - 52132131 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map