Assay Details
Target Gene Details
Entrez Gene ID: | 9212 |
Gene Name: | aurora kinase B |
Gene Aliases: |
AIK2, AIM-1, AIM1, ARK2, AurB, IPL1, PPP1R48, STK12, STK5, aurkb-sv1, aurkb-sv2 |
Location: |
Chr.17:8204731-8210768 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3639878 | Chr.17:8193720 - 8214825 on Build GRCh38 | Loss |
|
| nsv833353 | Chr.17:8150060 - 8326783 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map