Assay Details
Target Gene Details
Entrez Gene ID: | 170302 |
Gene Name: | aristaless related homeobox |
Gene Aliases: |
CT121, EIEE1, ISSX, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS |
Location: |
Chr.X:25003694-25015948 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARX | NM_139058.2 | 1 | 253 | NP_620689.1 |
| AY038071.1 | AAK93901.1 | |||
| BQ100952.1 | 1 | 253 |
More Information
Set Membership: |
|

Genomic Map