Assay Details
Target Gene Details
Entrez Gene ID: | 79651 |
Gene Name: | rhomboid 5 homolog 2 |
Gene Aliases: |
RHBDL5, RHBDL6, TEC, TOC, TOCG, iRhom2 |
Location: |
Chr.17:76470893-76501440 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RHBDF2 | NM_001005498.3 | NP_001005498.2 | ||
| NM_024599.5 | NP_078875.4 | |||
| XM_005257669.3 | XP_005257726.1 | |||
| XM_005257670.1 | XP_005257727.1 | |||
| XM_011525249.2 | XP_011523551.1 | |||
| XM_011525250.2 | XP_011523552.1 | |||
| XM_011525251.2 | XP_011523553.1 | |||
| XM_017025079.1 | XP_016880568.1 | |||
| AK025994.1 | BAB15310.1 | |||
| AK074034.1 | 16 | 2725 | BAB84860.1 | |
| AK225684.1 | ||||
| AK292135.1 | ||||
| AK292166.1 | ||||
| AL833680.1 | ||||
| AY500247.1 | AAS77567.1 | |||
| BC016034.1 | AAH16034.2 | |||
| BC035829.1 | AAH35829.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058263 | Chr.17:76466514 - 76530556 on Build GRCh38 | Gain |
|
| nsv833549 | Chr.17:76365444 - 76527890 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map