Assay Details
Target Gene Details
Entrez Gene ID: | 10269 |
Gene Name: | zinc metallopeptidase STE24 |
Gene Aliases: |
FACE-1, FACE1, HGPS, PRO1, STE24, Ste24p |
Location: |
Chr.1:40258050-40294184 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMPSTE24 | NM_005857.4 | 2 | 428 | NP_005848.2 |
| AB016068.1 | 2 | 384 | BAA33727.1 | |
| AF064867.1 | 2 | 329 | AAC68866.1 | |
| AK074784.1 | 2 | 284 | ||
| AK075007.1 | 2 | 307 | ||
| BC037283.1 | 2 | 332 | AAH37283.1 | |
| DA398037.1 | 2 | 400 | ||
| DB096663.1 | 2 | 430 | ||
| DB285015.1 | 2 | 417 | ||
| Y13834.1 | 2 | 239 | CAB46277.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829660 | Chr.1:40073032 - 40266408 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map