Assay Details
Target Gene Details
Entrez Gene ID: | 1410 |
Gene Name: | crystallin alpha B |
Gene Aliases: |
CMD1II, CRYA2, CTPP2, CTRCT16, HEL-S-101, HSPB5, MFM2 |
Location: |
Chr.11:111908620-111913213 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRYAB | NM_001289807.1 | NP_001276736.1 | ||
| NM_001885.2 | NP_001876.1 | |||
| BI759672.1 | ||||
| BM721803.1 | ||||
| DB506606.1 |
Target Gene Details
Entrez Gene ID: | 3316 |
Gene Name: | heat shock protein family B (small) member 2 |
Gene Aliases: |
HSP27, Hs.78846, LOH11CR1K, MKBP |
Location: |
Chr.11:111912736-111914093 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPB2 | NM_001541.3 | NP_001532.1 | ||
| AB096250.1 | ||||
| BC109393.1 | AAI09394.1 | |||
| CR457408.1 | CAG33689.1 | |||
| D89617.1 | BAA24004.1 |
Target Gene Details
Entrez Gene ID: | 100528019 |
Gene Name: | HSPB2-C11orf52 readthrough (NMD candidate) |
Gene Aliases: |
- |
Location: |
Chr.11:111912736-111926871 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HSPB2-C11orf52 | NR_037651.1 | |||
| AK293041.1 | ||||
| BC110872.1 | AAI10873.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1070987 | Chr.11:111912775 - 111913376 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map