Assay Details
Target Gene Details
Entrez Gene ID: | 9679 |
Gene Name: | family with sequence similarity 53 member B |
Gene Aliases: |
KIAA0140, bA12J10.2, smp |
Location: |
Chr.10:124619294-124744361 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM53B | NM_014661.3 | NP_055476.3 | ||
| AK127343.1 | ||||
| BC031654.1 | AAH31654.1 | |||
| BX344666.2 | ||||
| CA397342.1 | ||||
| D50930.1 |
Target Gene Details
Entrez Gene ID: | 101927944 |
Gene Name: | FAM53B antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.10:124704028-124714774 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM53B-AS1 | NR_120630.1 | |||
| NR_120631.1 | ||||
| AW003106.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv470973 | Chr.10:124658034 - 124736254 on Build GRCh38 | Loss |
|
| nsv832013 | Chr.10:124557573 - 124776461 on Build GRCh38 | Loss |
|
| esv3584565 | Chr.10:124637794 - 124707783 on Build GRCh38 | Gain |
|
| nsv521161 | Chr.10:124518637 - 124736254 on Build GRCh38 | Gain+Loss |
|
| nsv1050837 | Chr.10:124678954 - 124828763 on Build GRCh38 | Gain |
|
| nsv832014 | Chr.10:124674236 - 124862247 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map