Assay Details
Target Gene Details
Entrez Gene ID: | 64222 |
Gene Name: | torsin family 3 member A |
Gene Aliases: |
ADIR, ADIR2 |
Location: |
Chr.1:179081977-179095996 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR3A | NM_022371.3 | 3 | 690 | NP_071766.2 |
| AJ299403.1 | 3 | 579 | CAC13973.1 | |
| AJ299441.1 | 3 | 579 | CAC14461.1 | |
| AK025998.1 | 3 | 569 | BAB15312.1 | |
| AK130132.1 | ||||
| AK299966.1 | ||||
| AK316530.1 | ||||
| BC001085.1 | 3 | 588 | AAH01085.1 | |
| BC007571.1 | 3 | 579 | AAH07571.1 | |
| BC011746.1 | 3 | 575 | AAH11746.1 | |
| BC018292.1 | 3 | 580 | AAH18292.1 | |
| BC088368.1 | AAH88368.1 | |||
| CR457354.1 | 3 | 538 | CAG33635.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv3721 | Chr.1:179050736 - 179094159 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map