Assay Details
Target Gene Details
Entrez Gene ID: | 4319 |
Gene Name: | matrix metallopeptidase 10 |
Gene Aliases: |
SL-2, STMY2 |
Location: |
Chr.11:102770502-102780628 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP10 | NM_002425.2 | NP_002416.1 | ||
| AK222601.1 | BAD96321.1 | |||
| AK313960.1 | ||||
| BC002591.2 | AAH02591.1 | |||
| BT007442.1 | AAP36110.1 | |||
| X07820.1 | CAA30679.1 |
More Information
Set Membership: |
|

Genomic Map