Assay Details
Target Gene Details
Entrez Gene ID: | 196403 |
Gene Name: | deltex E3 ubiquitin ligase 3 |
Gene Aliases: |
RNF154, deltex3 |
Location: |
Chr.12:57604327-57609804 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DTX3 | NM_001286245.1 | 6 | 1934 | NP_001273174.1 |
| NM_001286246.1 | 6 | 1953 | NP_001273175.1 | |
| NM_178502.3 | 7 | 2114 | NP_848597.1 | |
| XM_005268697.1 | 7 | 2617 | XP_005268754.1 | |
| XM_005268698.1 | 7 | 2390 | XP_005268755.1 | |
| XM_005268700.1 | 7 | 2270 | XP_005268757.1 | |
| XM_005268703.1 | 7 | 2289 | XP_005268760.1 | |
| XM_011538022.2 | 6 | 2492 | XP_011536324.1 | |
| AK092085.1 | 7 | 1915 | BAC03801.1 | |
| AK094385.1 | 6 | 1723 | BAC04344.1 | |
| AK128752.1 | 5 | 3082 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559034 | Chr.12:57466149 - 57618450 on Build GRCh38 | Loss |
|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map