Assay Details
Target Gene Details
Entrez Gene ID: | 23676 |
Gene Name: | small muscle protein, X-linked |
Gene Aliases: |
DFN6, DFNX4 |
Location: |
Chr.X:21705972-21758160 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMPX | NM_014332.2 | 5 | 741 | NP_055147.1 |
| NR_045617.1 | 6 | 843 | ||
| AF129505.1 | 5 | 693 | AAF19343.1 | |
| AJ250584.1 | 5 | 693 | CAC08492.1 | |
| BC005948.1 | 5 | 622 | AAH05948.1 | |
| BF126275.1 | ||||
| BM674432.1 | 1 | 211 |
More Information
Set Membership: |
|

Genomic Map