Assay Details
Target Gene Details
Entrez Gene ID: | 114826 |
Gene Name: | SET and MYND domain containing 4 |
Gene Aliases: |
ZMYND21 |
Location: |
Chr.17:1779535-1829896 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMYD4 | NM_052928.2 | 8 | 2108 | NP_443160.2 |
| XM_011523632.2 | 8 | 2089 | XP_011521934.1 | |
| XM_011523633.2 | 9 | 2109 | XP_011521935.1 | |
| XM_017024121.1 | 9 | 2353 | XP_016879610.1 | |
| AB067523.1 | 4 | 1352 | BAB67829.1 | |
| AK057769.1 | 4 | 540 | BAB71564.1 | |
| AK092149.1 | 4 | 1013 | ||
| AK095369.1 | 8 | 2108 | BAC04538.1 | |
| BC035077.2 | 8 | 2027 | AAH35077.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv9490 | Chr.17:1653428 - 1828993 on Build GRCh38 | Loss |
|
| esv33335 | Chr.17:1779713 - 2048672 on Build GRCh38 | Loss |
|
| nsv1065585 | Chr.17:1733183 - 1918769 on Build GRCh38 | Gain |
|
| nsv833335 | Chr.17:1713955 - 1827844 on Build GRCh38 | Gain+Loss |
|
| nsv574215 | Chr.17:1779718 - 1824004 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map