Assay Details
Target Gene Details
Entrez Gene ID: | 57761 |
Gene Name: | tribbles pseudokinase 3 |
Gene Aliases: |
C20orf97, NIPK, SINK, SKIP3, TRB3 |
Location: |
Chr.20:380629-397559 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIB3 | NM_001301188.1 | 4 | 676 | NP_001288117.1 |
| NM_001301190.1 | 4 | 691 | NP_001288119.1 | |
| NM_001301193.1 | 4 | 789 | NP_001288122.1 | |
| NM_001301196.1 | 4 | 848 | NP_001288125.1 | |
| NM_001301201.1 | 5 | 1042 | NP_001288130.1 | |
| NM_021158.4 | 4 | 1183 | NP_066981.2 | |
| XM_017027989.1 | 4 | 1167 | XP_016883478.1 | |
| AF250311.1 | 3 | 648 | AAK58175.1 | |
| AJ697936.1 | 5 | 1026 | CAG27047.1 | |
| AK026945.1 | 5 | 936 | BAB15597.1 | |
| AK222837.1 | 5 | 936 | BAD96557.1 | |
| AK297546.1 | 5 | 1040 | ||
| AY247738.1 | 4 | 1182 | AAP04407.1 | |
| BC019363.1 | 4 | 749 | AAH19363.1 | |
| BC027484.1 | 4 | 936 | AAH27484.1 | |
| BG828172.1 | ||||
| CR457366.1 | 3 | 642 | CAG33647.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3644951 | Chr.20:385214 - 399104 on Build GRCh38 | Loss |
|
| esv3644952 | Chr.20:386013 - 1119183 on Build GRCh38 | Gain |
|
| nsv510777 | Chr.20:366849 - 416704 on Build GRCh38 | Deletion |
|
| nsv585170 | Chr.20:380145 - 422066 on Build GRCh38 | Loss |
|
| esv2657243 | Chr.20:385360 - 399238 on Build GRCh38 | Deletion |
|
| nsv519415 | Chr.20:389506 - 396582 on Build GRCh38 | Loss |
|
| dgv4219n100 | Chr.20:80652 - 461103 on Build GRCh38 | Gain |
|
| esv3893263 | Chr.20:216521 - 434561 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map