Assay Details
Target Gene Details
Entrez Gene ID: | 713 |
Gene Name: | complement C1q B chain |
Gene Aliases: |
- |
Location: |
Chr.1:22653189-22661536 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1QB | NM_000491.3 | 3 | 514 | NP_000482.3 |
| XM_011542059.2 | 4 | 665 | XP_011540361.1 | |
| BC008983.1 | 3 | 456 | AAH08983.1 | |
| BQ711602.1 | 4 | 676 | ||
| X03084.1 | 2 | 308 | CAA26880.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34161 | Chr.1:22599762 - 22817730 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map