Assay Details
Target Gene Details
Entrez Gene ID: | 29765 |
Gene Name: | tropomodulin 4 |
Gene Aliases: |
SK-TMOD |
Location: |
Chr.1:151169986-151176071 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMOD4 | NM_013353.2 | 8 | 1002 | NP_037485.2 |
| XM_011509449.1 | 8 | 884 | XP_011507751.1 | |
| XM_017001089.1 | 7 | 1619 | XP_016856578.1 | |
| XM_017001090.1 | XP_016856579.1 | |||
| AF165217.1 | 8 | 879 | AAF01277.1 | |
| AF177173.2 | 8 | 947 | AAF31672.1 | |
| AK092700.1 | 8 | 864 | BAC03951.1 | |
| AK300675.1 | 6 | 705 | ||
| AK313280.1 | 8 | 911 | ||
| BC017810.1 | 8 | 912 | AAH17810.1 | |
| BX647346.1 | 6 | 626 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522840 | Chr.1:150961247 - 151375270 on Build GRCh38 | Gain |
|
| dgv2n68 | Chr.1:151136983 - 151309302 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map