Assay Details
Target Gene Details
Entrez Gene ID: | 162605 |
Gene Name: | keratin 28 |
Gene Aliases: |
K25IRS4, KRT25D |
Location: |
Chr.17:40792196-40799959 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT28 | NM_181535.3 | NP_853513.2 | ||
| AJ564207.1 | CAD91907.1 | |||
| AK129827.1 | BAC85240.1 |
More Information
Set Membership: |
|

Genomic Map