Assay Details
Target Gene Details
Entrez Gene ID: | 3755 |
Gene Name: | potassium voltage-gated channel modifier subfamily G member 1 |
Gene Aliases: |
K13, KCNG, KV6.1, kH2 |
Location: |
Chr.20:51003656-51023176 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNG1 | NM_002237.3 | NP_002228.2 | ||
| XM_006723785.2 | XP_006723848.1 | |||
| XM_011528800.1 | XP_011527102.1 | |||
| XM_011528801.1 | XP_011527103.1 | |||
| XM_011528802.1 | XP_011527104.1 | |||
| XM_011528803.2 | XP_011527105.1 | |||
| XM_011528804.1 | XP_011527106.1 | |||
| XM_011528805.2 | XP_011527107.1 | |||
| XM_011528806.1 | XP_011527108.1 | |||
| AF033383.1 | AAC05635.1 | |||
| AK290689.1 | ||||
| BC006367.1 | 3 | 1734 | AAH06367.1 | |
| BC046629.2 | AAH46629.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv32899 | Chr.20:51009144 - 51012538 on Build GRCh38 | Gain |
|
| esv33244 | Chr.20:51009054 - 51009144 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map