Assay Details
Target Gene Details
Entrez Gene ID: | 7421 |
Gene Name: | vitamin D (1,25- dihydroxyvitamin D3) receptor |
Gene Aliases: |
NR1I1, PPP1R163 |
Location: |
Chr.12:47841537-47905031 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VDR | NM_000376.2 | 10 | 2999 | NP_000367.1 |
| NM_001017535.1 | 11 | 3121 | NP_001017535.1 | |
| NM_001017536.1 | 10 | 3390 | NP_001017536.1 | |
| AK024830.1 | 1 | 1123 | ||
| AK225844.1 | 10 | 2998 | ||
| BC060832.1 | 10 | 3362 | AAH60832.1 | |
| J03258.1 | 10 | 2954 | AAA61273.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs115591719] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map