Assay Details
Target Gene Details
Entrez Gene ID: | 7703 |
Gene Name: | polycomb group ring finger 2 |
Gene Aliases: |
MEL-18, RNF110, ZNF144 |
Location: |
Chr.17:38733897-38749817 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCGF2 | NM_007144.2 | NP_009075.1 | ||
| XM_005257640.2 | XP_005257697.1 | |||
| XM_005257641.4 | XP_005257698.1 | |||
| XM_005257642.3 | XP_005257699.1 | |||
| XM_017025016.1 | XP_016880505.1 | |||
| BC004858.2 | AAH04858.1 | |||
| BC024255.1 | AAH24255.1 | |||
| BX647429.1 | ||||
| D13969.1 | BAA03074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833438 | Chr.17:38593441 - 38821266 on Build GRCh38 | Loss |
|
| nsv470585 | Chr.17:38701569 - 38760346 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map